ENA413165834 Enamine

tert-butyl N-[(1R)-1-carbamoylpropyl]carbamate

  • MDL number MFCD26936057
  • Cas Number : 1807941-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139901
Availability details for
Quantity
  • Availability
  • Add to Cart