ENA413165835 Enamine

3-amino-N-(2-aminoethyl)-N-({[1,1'-biphenyl]-4-yl}methyl)propane-1-sulfonamide

  • MDL number MFCD27500693
  • Cas Number : 1803592-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139964
Availability details for
Quantity
  • Availability
  • Add to Cart