ENA413165839 Enamine

tert-butyl N-[(4-carbamoylfuran-2-yl)methyl]carbamate

  • MDL number MFCD27500694
  • Cas Number : 1803587-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140081
Availability details for
Quantity
  • Availability
  • Add to Cart