ENA413165841 Enamine

1-[4-({2-[(2-methylprop-2-en-1-yl)oxy]ethyl}amino)phenyl]ethan-1-one

  • MDL number MFCD27500695
  • Cas Number : 1707756-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140144
Availability details for
Quantity
  • Availability
  • Add to Cart