ENA413165842 Enamine

(2S)-2-amino-3-methylbutanenitrile hydrochloride

  • MDL number MFCD26936065
  • Cas Number : 1798904-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140211
Availability details for
Quantity
  • Availability
  • Add to Cart