ENA413165845 Enamine

tert-butyl N-[1-(1H-imidazol-2-yl)propyl]carbamate

  • MDL number MFCD26936072
  • Cas Number : 1595764-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140313

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140313
Availability details for
Quantity
  • Availability
  • Add to Cart