ENA413165848 Enamine

tert-butyl 3-(1-ethoxy-1-oxopropan-2-yl)azetidine-1-carboxylate

  • MDL number MFCD26936077
  • Cas Number : 1803607-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140353

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140353
Availability details for
Quantity
  • Availability
  • Add to Cart