ENA413165862 Enamine

5-[4-(5-chloro-2-methoxybenzenesulfonyl)piperazin-1-yl]-5-oxopentanoic acid

  • MDL number MFCD27500698
  • Cas Number : 1803580-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140457
Availability details for
Quantity
  • Availability
  • Add to Cart