ENA413165872 Enamine

2-[(cyclopropylmethyl)amino]acetic acid hydrochloride

  • MDL number MFCD26936109
  • Cas Number : 1803587-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201...203
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140576

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140576
Availability details for
Quantity
  • Availability
  • Add to Cart