ENA413165876 Enamine

5-{[(2-chloropyridin-4-yl)amino]methylidene}-2,2-dimethyl-1,3-dioxane-4,6-dione

  • MDL number MFCD26936115
  • Cas Number : 1018812-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205...207
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140615

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140615
Availability details for
Quantity
  • Availability
  • Add to Cart