ENA413165877 Enamine

(3-amino-1,1,1-trifluoropropan-2-yl)(benzyl)methylamine hydrochloride

  • MDL number MFCD26936116
  • Cas Number : 1803598-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140619

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140619
Availability details for
Quantity
  • Availability
  • Add to Cart