ENA413165878 Enamine

5-(4-fluorophenyl)pentanamide

  • MDL number MFCD27500701
  • Cas Number : 1798712-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-140622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140622
Availability details for
Quantity
  • Availability
  • Add to Cart