ENA413165879 Enamine

4-chloro-2-methyl-6-(trifluoromethyl)aniline hydrochloride

  • MDL number MFCD26936117
  • Cas Number : 1803566-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140632
Availability details for
Quantity
  • Availability
  • Add to Cart