ENA413165882 Enamine

ethyl 3-(4-fluorophenyl)-2-methylpropanoate

  • MDL number MFCD26936123
  • Cas Number : 1803591-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140699

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140699
Availability details for
Quantity
  • Availability
  • Add to Cart