ENA413165885 Enamine

2-(6-fluoro-1-methyl-1H-indol-3-yl)ethan-1-amine hydrochloride

  • MDL number MFCD26936127
  • Cas Number : 1689575-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140741

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140741
Availability details for
Quantity
  • Availability
  • Add to Cart