ENA413165887 Enamine

2-[4-(aminomethyl)-2-chloro-6-methoxyphenoxy]acetamide hydrochloride

  • MDL number MFCD26936134
  • Cas Number : 1803600-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230...232
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140780

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140780
Availability details for
Quantity
  • Availability
  • Add to Cart