ENA413165893 Enamine

methyl 5-(fluorosulfonyl)-3-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD26936141

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140865
Availability details for
Quantity
  • Availability
  • Add to Cart