ENA413165897 Enamine

N'-(2-chloropropanoyl)-2-fluorobenzohydrazide

  • MDL number MFCD26936145
  • Cas Number : 1803584-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140899

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140899
Availability details for
Quantity
  • Availability
  • Add to Cart