ENA413165898 Enamine

2-{1-[5-(2-fluorophenyl)-1,3,4-thiadiazol-2-yl]ethyl}-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD26936146
  • Cas Number : 1795518-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140900
Availability details for
Quantity
  • Availability
  • Add to Cart