ENA413165900 Enamine

(2,2-difluoroethyl)(methyl)[2-(methylamino)ethyl]amine dihydrochloride

  • MDL number MFCD26936149
  • Cas Number : 1803580-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140954
Availability details for
Quantity
  • Availability
  • Add to Cart