ENA413165903 Enamine

(4-fluoro-3-nitrophenyl)hydrazine hydrochloride

  • MDL number MFCD26936156
  • Cas Number : 1033439-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141029

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141029
Availability details for
Quantity
  • Availability
  • Add to Cart