ENA413165905 Enamine

2-(cyclopropylmethoxy)ethyl methanesulfonate

  • MDL number MFCD26936158
  • Cas Number : 63659-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141040
Availability details for
Quantity
  • Availability
  • Add to Cart