ENA413165911 Enamine

tert-butyl N-[3-(2-chloro-6-methylphenyl)prop-2-yn-1-yl]carbamate

  • MDL number MFCD26936167
  • Cas Number : 1803580-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141123
Availability details for
Quantity
  • Availability
  • Add to Cart