ENA413165928 Enamine

ethyl 2-amino-2-(2,3-dimethylphenyl)acetate hydrochloride

  • MDL number MFCD26936185
  • Cas Number : 1803572-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141446

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141446
Availability details for
Quantity
  • Availability
  • Add to Cart