ENA413165931 Enamine

2-amino-N-[(1-cyanocyclopropyl)methyl]acetamide hydrochloride

  • MDL number MFCD26936189
  • Cas Number : 1803609-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141464
Availability details for
Quantity
  • Availability
  • Add to Cart