ENA413165936 Enamine

4,4-dimethylcyclohexyl methanesulfonate

  • MDL number MFCD26936196
  • Cas Number : 1449214-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141605
Availability details for
Quantity
  • Availability
  • Add to Cart