ENA413165937 Enamine

(3-methoxyphenyl)(pyridin-2-yl)methanamine dihydrochloride

  • MDL number MFCD26936197
  • Cas Number : 1803595-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141611
Availability details for
Quantity
  • Availability
  • Add to Cart