ENA413165938 Enamine

2,2-dimethyl-3-(methylamino)propanoic acid hydrochloride

  • MDL number MFCD26936200
  • Cas Number : 141950-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155...157
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141630

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141630
Availability details for
Quantity
  • Availability
  • Add to Cart