ENA413165939 Enamine

3-[(thiophen-3-yl)methyl]-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride

  • MDL number MFCD26936201
  • Cas Number : 1803610-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141637
Availability details for
Quantity
  • Availability
  • Add to Cart