ENA413165940 Enamine

3,6-dimethylpyridazin-4-amine hydrochloride

  • MDL number MFCD26936202
  • Cas Number : 1803610-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141639
Availability details for
Quantity
  • Availability
  • Add to Cart