ENA413165942 Enamine

4-bromo-2-tert-butyl-6-nitrophenol

  • MDL number MFCD27500713
  • Cas Number : 150313-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141689
Availability details for
Quantity
  • Availability
  • Add to Cart