ENA413165943 Enamine

2-{1-[(tert-butoxy)carbonyl]azetidin-3-ylidene}propanoic acid

  • MDL number MFCD26936208
  • Cas Number : 1593231-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141703

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141703
Availability details for
Quantity
  • Availability
  • Add to Cart