ENA413165955 Enamine

5-(difluoromethoxy)-8-nitroquinoline

  • MDL number MFCD26936222
  • Cas Number : 1803585-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141834

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141834
Availability details for
Quantity
  • Availability
  • Add to Cart