ENA413165956 Enamine

1,3-diethyl 2-(1-phenylcyclopentanecarbonyl)propanedioate

  • MDL number MFCD26936223
  • Cas Number : 1803584-37-3
  • Molecular Weight : 332.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141846
Availability details for
Quantity
  • Availability
  • Add to Cart