ENA413165957 Enamine

4-(1H-pyrrol-1-yl)piperidine hydrochloride

  • MDL number MFCD26936224
  • Cas Number : 1803570-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141875
Availability details for
Quantity
  • Availability
  • Add to Cart