ENA413165962 Enamine

2-cyclopropanecarbonylmorpholine; trifluoroacetic acid

  • MDL number MFCD26936234
  • Cas Number : 1803585-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141947

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141947
Availability details for
Quantity
  • Availability
  • Add to Cart