ENA413165965 Enamine

N-methyl-3,4-dihydro-2H-pyrrol-5-amine hydrochloride

  • MDL number MFCD26936239
  • Cas Number : 7544-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142019

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142019
Availability details for
Quantity
  • Availability
  • Add to Cart