ENA413165967 Enamine

3-tert-butyl-5-(trichloromethyl)-1,2,4-thiadiazole

  • MDL number MFCD27920054
  • Cas Number : 1803586-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142040
Availability details for
Quantity
  • Availability
  • Add to Cart