ENA413165968 Enamine

1-(4-fluorobenzoyl)cyclopropan-1-amine hydrochloride

  • MDL number MFCD26936240
  • Cas Number : 1803592-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142045
Availability details for
Quantity
  • Availability
  • Add to Cart