ENA413165969 Enamine

3-tert-butyl-1-[(pyridin-4-yl)methyl]-1H-pyrazol-5-amine

  • MDL number MFCD26936241
  • Cas Number : 1803611-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-142053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142053
Availability details for
Quantity
  • Availability
  • Add to Cart