ENA413165975 Enamine

4-bromooxane-4-carbonyl chloride

  • MDL number MFCD27500719
  • Cas Number : 1803591-91-4
  • Molecular Weight : 227.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142108

Safety Information

signalword Danger
pcodes P234,P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P390,P403+P233,P405,P406,P501
MSDS https://www.enaminestore.com/catalog/EN300-142108
pictograms GHS05,GHS07
hstatements H290,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart