ENA413165977 Enamine

2-(cyclohex-1-en-1-yl)-2-methylpropan-1-amine

  • MDL number MFCD26936251
  • Cas Number : 1598255-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142145
Availability details for
Quantity
  • Availability
  • Add to Cart