ENA413165981 Enamine

2,5-dimethyl-1-[4-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD27920056
  • Cas Number : 1803580-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142188
Availability details for
Quantity
  • Availability
  • Add to Cart