ENA413165983 Enamine

5-bromo-8-chloro-1,2,3,4-tetrahydroquinoline hydrochloride

  • MDL number MFCD26936256
  • Cas Number : 1803607-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142239

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142239
Availability details for
Quantity
  • Availability
  • Add to Cart