ENA413165984 Enamine

methyl 4-bromooxane-4-carboxylate

  • MDL number MFCD26936257
  • Cas Number : 1192067-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142245

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142245
Availability details for
Quantity
  • Availability
  • Add to Cart