ENA413165990 Enamine

2-({[4-(3-bromophenyl)-1,3-thiazol-2-yl]methyl}amino)ethan-1-ol

  • MDL number MFCD26936264
  • Cas Number : 1803607-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-142352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142352
Availability details for
Quantity
  • Availability
  • Add to Cart