ENA413165991 Enamine

1',2'-dihydrospiro[cyclopropane-1,3'-indole] hydrochloride

  • MDL number MFCD26936267
  • Cas Number : 1788041-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142356

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-142356
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart