ENA413165999 Enamine

N-(azetidin-3-yl)cyclopropanecarboxamide hydrochloride

  • MDL number MFCD26936275
  • Cas Number : 1803599-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142432
Availability details for
Quantity
  • Availability
  • Add to Cart