ENA413166006 Enamine

4-[(4-fluorophenyl)methyl]piperidin-4-ol hydrochloride

  • MDL number MFCD26936282
  • Cas Number : 1429505-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224...226
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142543

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142543
Availability details for
Quantity
  • Availability
  • Add to Cart