ENA413166007 Enamine

methyl 3-methylpiperidine-4-carboxylate hydrochloride

  • MDL number MFCD27500726
  • Cas Number : 1797255-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142581

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142581
Availability details for
Quantity
  • Availability
  • Add to Cart